Merck
A Human Pluripotent Stem Cell-based Platform to Study SARS-CoV-2 Tropism and Model Virus Infection in Human Cells and Organoids.
Cell stem cell (2020-06-25)
Liuliu Yang, Yuling Han, Benjamin E Nilsson-Payant, Vikas Gupta, Pengfei Wang, Xiaohua Duan, Xuming Tang, Jiajun Zhu, Zeping Zhao, Fabrice Jaffré, Tuo Zhang, Tae Wan Kim, Oliver Harschnitz, David Redmond, Sean Houghton, Chengyang Liu, Ali Naji, Gabriele Ciceri, Sudha Guttikonda, Yaron Bram, Duc-Huy T Nguyen, Michele Cioffi, Vasuretha Chandar, Daisy A Hoagland, Yaoxing Huang, Jenny Xiang, Hui Wang, David Lyden, Alain Borczuk, Huanhuan Joyce Chen, Lorenz Studer, Fong Cheng Pan, David D Ho, Benjamin R tenOever, Todd Evans, Robert E Schwartz, Shuibing Chen
ABSTRAKT
SARS-CoV-2 has caused the COVID-19 pandemic. There is an urgent need for physiological models to study SARS-CoV-2 infection using human disease-relevant cells. COVID-19 pathophysiology includes respiratory failure but involves other organ systems including gut, liver, heart, and pancreas. We present an experimental platform comprised of cell and organoid derivatives from human pluripotent stem cells (hPSCs). A Spike-enabled pseudo-entry virus infects pancreatic endocrine cells, liver organoids, cardiomyocytes, and dopaminergic neurons. Recent clinical studies show a strong association with COVID-19 and diabetes. We find that human pancreatic beta cells and liver organoids are highly permissive to SARS-CoV-2 infection, further validated using adult primary human islets and adult hepatocyte and cholangiocyte organoids. SARS-CoV-2 infection caused striking expression of chemokines, as also seen in primary human COVID-19 pulmonary autopsy samples. hPSC-derived cells/organoids provide valuable models for understanding the cellular responses of human tissues to SARS-CoV-2 infection and for disease modeling of COVID-19.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
SANT-1, ≥98% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
8-Bromoadenosine 3′,5′-cyclic monophosphate, ≥97% (HPLC)
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
Sigma-Aldrich
γ-Secretase Inhibitor XX, This γ-secretase inhibitor, CAS 209984-56-5, is a cell-permeable dibenzazepine compound that lowers both brain and plasma Aβ40 levels by ~72% in Tg2576 mutant APP transgenic mouse model.