Merck
Crystal structures of Helicobacter pylori type II dehydroquinase inhibitor complexes: new directions for inhibitor design.
Journal of medicinal chemistry (2006-02-17)
David A Robinson, Kirsty A Stewart, Nicholas C Price, Peter A Chalk, John R Coggins, Adrian J Lapthorn
ABSTRAKT
The crystal structures of the type II dehydroquinase (DHQase) from Helicobacter pylori in complex with three competitive inhibitors have been determined. The inhibitors are the substrate analogue 2,3-anhydroquinate (FA1), citrate, and an oxoxanthene sulfonamide derivative (AH9095). Despite the very different chemical nature of the inhibitors, in each case the primary point of interaction with the enzyme is via the residues that bind the C1 functionalities of the substrate, 3-dehydroquinate, i.e., N76, H102, I103, and H104. The DHQase/AH9095 complex crystal structure shows that sulfonamides can form a scaffold for nonsubstrate-like inhibitors and identifies a large conserved hydrophobic patch at the entrance to the active site as a locus that can be exploited in the development of new ligands.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Citric acid, 99%