Merck
  • Metal-mediated inhibition of Escherichia coli methionine aminopeptidase: structure-activity relationships and development of a novel scoring function for metal-ligand interactions.
Metal-mediated inhibition of Escherichia coli methionine aminopeptidase: structure-activity relationships and development of a novel scoring function for metal-ligand interactions.
Journal of medicinal chemistry (2006-01-20)
Rolf Schiffmann, Alexander Neugebauer, Christian D Klein
ABSTRAKT
We report the discovery of thiabendazole as a potent inhibitor (K(i) = 0.4 microM) of Escherichia coli methionine aminopeptidase (ecMetAP) and the synthesis and pharmacological evaluation of thiabendazole congeners with activity in the upper nanomolar range. Elucidation of the X-ray structure of ecMetAP in complex with thiabendazole and an unrelated inhibitor that was independently described by another group showed that that both compounds bind to an additional Co(II) ion at the entrance of the active site. This unexpected finding explains the inactivity of the compounds under in vivo conditions. It also allows us to discuss the structure-activity relationships of this series of compounds in a meaningful way, based upon docking runs with an auxiliary metal ion. We describe a new scoring function for the evaluation of metal-mediated inhibitor binding that, unlike the previously used scoring function implemented in the docking program, allows us to distinguish between active and inactive compounds. Finally, conclusions for the structure-based design of in vivo-active inhibitors of ecMetAP are drawn.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Albendazole, analytical standard, ≥98%
Sigma-Aldrich
Thiabendazole, BioReagent, suitable for plant cell culture, powder
Supelco
Thiabendazole, PESTANAL®, analytical standard
Sigma-Aldrich
Thiabendazole, ≥99%, powder
Sigma-Aldrich
Fenbendazole, ≥98%
Supelco
Mebendazole, VETRANAL®, analytical standard
Supelco
Mebendazole, analytical standard, ≥98% (HPLC)
Sigma-Aldrich
Carbendazim, 97%
Supelco
Carbendazim, PESTANAL®, analytical standard