Merck
Spasmolytic effects of nonprenylated rotenoid constituents of Boerhaavia diffusa roots.
Journal of natural products (2006-06-24)
Francesca Borrelli, Valeria Ascione, Raffaele Capasso, Angelo A Izzo, Ernesto Fattorusso, Orazio Taglialatela-Scafati
ABSTRAKT
Boerhaavia diffusa is an Ayurvedic remedy used traditionally for the treatment of a number of diseases, including those affecting the gastrointestinal tract. In the current investigation, a methanol extract obtained from roots of B. diffusa exhibited a significant spasmolytic activity in the guinea pig ileum, probably through a direct effect on the smooth muscle. A detailed phytochemical analysis of this methanol extract led to the isolation of one new (12) and six known (6-11) rotenoid derivatives. The structure of the new compound was determined through interpretation of its MS and NMR data. All the isolated rotenoids were evaluated for their effect on intestinal motility in vitro, and the results obtained showed unambiguously that they are active spasmolytic constituents. Preliminary structure-activity relationships for this class of compounds are suggested.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)