Merck
Lysophosphatidic Acid and IL-6 Trans-signaling Interact via YAP/TAZ and STAT3 Signaling Pathways in Human Trabecular Meshwork Cells.
Investigative ophthalmology & visual science (2020-11-21)
Felix Yemanyi, VijayKrishna Raghunathan
ABSTRAKT
Lysophosphatidic acid (LPA) and soluble interleukin-6 receptor (sIL6R) are elevated in primary open angle glaucoma (POAG). LPA and IL6 modulate in response to biomechanical stimuli and converge on similar fibrotic phenotypes. Thus, we determined whether LPA and IL6 trans-signaling (IL6/sIL6R) interact via Yes-associated protein (YAP)/Transcriptional coactivator with a PDZ-binding motif (TAZ) or Signal transducer and activator of transcription 3 (STAT3) pathways in human trabecular meshwork (hTM) cells. Confluent primary hTM cells were serum starved for 24 hours, and treated with vehicle, LPA (20 µM), IL6 (100 ng/mL)/sIL6R (200 ng/mL), or both (LPA + IL6/sIL6R) for 24 hours, with or without a YAP inhibitor (verteporfin; 2 µM) or STAT3 inhibitor (2 µM). Expression of key receptors and ligands, signaling mediators, actomyosin machinery, cell contractility, and extracellular matrix (ECM) targets of both signaling pathways was determined by immunocytochemistry, RT-qPCR, and Western blotting. LPA and IL6 trans-signaling coupling overexpressed/activated receptors and ligands, glycoprotein-130, IL6, and autotaxin; signaling mediators, YAP, TAZ, Pan-TEAD, and phosphorylated STAT3 (pSTAT3); actomyosin and contractile machinery components, myosin light chain 2 (MLC2), phosphorylated MLC2, rho-associated protein kinase 1, filamentous actin, and α-smooth muscle actin; and fibrotic ECM proteins, collagen I and IV, fibronectin, laminin, cysteine-rich angiogenic inducer 61, and connective tissue growth factor in hTM cells; mostly beyond LPA or IL6 trans-signaling alone. Verteporfin inhibited YAP, TAZ, and pSTAT3, with concomitant abrogation of aforementioned fibrotic targets; the STAT3 inhibitor was only partially effective. These data suggest synergistic crosstalk between LPA and IL6 trans-signaling, mediated by YAP, TAZ, and pSTAT3. By completely inhibiting these mediators, verteporfin may be more efficacious in ameliorating LPA and/or IL6 trans-signaling-induced ocular hypertensive phenotypes in hTM cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
STAT3 Inhibitor III, WP1066, STAT3 Inhibitor III, WP1066, CAS 857064-38-1, is a cell-permeable AG490 tyrphostin analog that acts as a STAT3 pathway inhibitor. A potent anti-tumor agent.
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
sIL-6 Receptor alpha human, recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.