Merck
Haematopoietic stem cell-dependent Notch transcription is mediated by p53 through the Histone chaperone Supt16h.
Nature cell biology (2020-11-25)
Sophia G Espanola, Hyemin Song, Eunjin Ryu, Aditya Saxena, Eun-Sun Kim, Jennifer E Manegold, Chanond A Nasamran, Debashis Sahoo, Chang-Kyu Oh, Cara Bickers, Unbeom Shin, Stephanie Grainger, Yong Hwan Park, Lauren Pandolfo, Mi-Sun Kang, Sukhyun Kang, Kyungjae Myung, Kimberly L Cooper, Deborah Yelon, David Traver, Yoonsung Lee
ABSTRAKT
Haematopoietic stem and progenitor cells (HSPCs) have been the focus of developmental and regenerative studies, yet our understanding of the signalling events regulating their specification remains incomplete. We demonstrate that supt16h, a component of the Facilitates chromatin transcription (FACT) complex, is required for HSPC formation. Zebrafish supt16h mutants express reduced levels of Notch-signalling components, genes essential for HSPC development, due to abrogated transcription. Whereas global chromatin accessibility in supt16h mutants is not substantially altered, we observe a specific increase in p53 accessibility, causing an accumulation of p53. We further demonstrate that p53 influences expression of the Polycomb-group protein PHC1, which functions as a transcriptional repressor of Notch genes. Suppression of phc1 or its upstream regulator, p53, rescues the loss of both Notch and HSPC phenotypes in supt16h mutants. Our results highlight a relationship between supt16h, p53 and phc1 to specify HSPCs via modulation of Notch signalling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Di(N-succinimidyl) glutarate, ≥97.0% (CHN)
Informacje o cenach i dostępności nie są obecnie dostępne.