Merck
  • 2-Aminopropane-1,2,3-tricarboxylic acid: Synthesis and co-crystallization with the class A beta-lactamase BS3 of Bacillus licheniformis.
2-Aminopropane-1,2,3-tricarboxylic acid: Synthesis and co-crystallization with the class A beta-lactamase BS3 of Bacillus licheniformis.
Bioorganic & medicinal chemistry letters (2008-06-03)
Joséphine Beck, Eric Sauvage, Paulette Charlier, Jacqueline Marchand-Brynaert
ABSTRAKT
The title compound 4 has been prepared in four steps from ethylglycinate in 63% overall yield. This amino analog of citric acid has been co-crystallized with the class A beta-lactamase BS3 of Bacillus licheniformis and the structure of the complex fully analyzed by X-ray diffraction. Tris-ethyl aminocitrate 3 and the free tris-acid 4 have been tested against a member beta-lactamase from all distinct subgroups. They are novel inhibitors of class A beta-lactamases, still modest but more potent than citrate and isocitrate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Citric acid, 99%