Merck
Palmitoylation-dependent endosomal localization of AATYK1A and its interaction with Src.
Genes to cells : devoted to molecular & cellular mechanisms (2008-08-12)
Koji Tsutsumi, Mineko Tomomura, Teiichi Furuichi, Shin-ichi Hisanaga
ABSTRAKT
Apoptosis-associated tyrosine kinase 1 (AATYK1), also named LMTK1, was previously isolated as an apoptosis-related gene from 32Dcl3 myeloid precursor cells, but its precise function remains unknown. AATYK1A, an isoform without a transmembrane domain, is highly expressed in neurons. We identified palmitoylation of AATYK1A at three N-terminal cysteine residues in cortical cultured neurons and COS-7 cells and found that palmitoylation determined localization of AATYK1A to the transferrin receptor-positive recycling endosomes. Further, we identified the tyrosine kinase Src as a novel AATYK1A-interacting protein. Src and Fyn phosphorylated AATYK1A at tyrosines 25 and 46 in a palmitoylation-dependent manner. The association of AATYK1A with Src in endosomes was also found to be palmitoylation-dependent. These results indicate that palmitoylation is a critical factor not only for the subcellular localization of AATYK1A but also for its interaction with Src.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało przeciw fosfotyrozynie, klon ®, clone 4G10®, Upstate®, from mouse
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Przeciwciało anty-V-Src, klon 327, clone 327, from mouse
Informacje o cenach i dostępności nie są obecnie dostępne.