Merck
  • 1-Methyl and 1-(2-hydroxyalkyl)-5-(3-alkyl/cycloalkyl/phenyl/naphthylureido)-1H-pyrazole-4-carboxylic acid ethyl esters as potent human neutrophil chemotaxis inhibitors.
1-Methyl and 1-(2-hydroxyalkyl)-5-(3-alkyl/cycloalkyl/phenyl/naphthylureido)-1H-pyrazole-4-carboxylic acid ethyl esters as potent human neutrophil chemotaxis inhibitors.
Bioorganic & medicinal chemistry (2009-04-14)
Olga Bruno, Chiara Brullo, Francesco Bondavalli, Silvia Schenone, Susanna Spisani, Maria Sofia Falzarano, Katia Varani, Elisabetta Barocelli, Vigilio Ballabeni, Carmine Giorgio, Massimiliano Tognolini
ABSTRAKT
In this paper we report the synthesis and the chemotaxis inhibitory activity of a number of 1H-pyrazole-4-carboxylic acid ethyl esters 2 functionalized in N1 with a methyl group or different hydroxyalkyl chains and in position 5 with a series of 3-substituted urea groups. These compounds were designed as development of previous pyrazole-urea derivatives that resulted potent IL8-induced neutrophil chemotaxis inhibitors in vitro. Most of the new compounds revealed a potent inhibition of both IL8- and fMLP-OMe-stimulated neutrophil chemotaxis. The most active compounds in the fMLP-OMe induced chemotaxis test showed IC(50) in the range 0.19 nM-2 microM; but we observed a very strong inhibition in the IL8-induced chemotaxis test, having the most active compounds IC(50) at pM concentrations. In vivo compounds 2e and 2f, although to a lesser extent, at 50mg/kg os decreased granulocyte infiltration in zymosan-induced peritonitis in mice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid