Merck
The endothelial nitric oxide synthase/cyclic guanosine monophosphate/protein kinase G pathway activates primordial follicles.
Aging (2020-12-09)
Peikun Zhao, Zidai Song, Yan Wang, Han Cai, Xiaoyan Du, Changlong Li, Jianyi Lv, Xin Liu, Meng Guo, Zhenwen Chen
ABSTRAKT
In mammals, the well-organized activation of quiescent primordial follicles is pivotal for female reproductive reserve. In the present study, we examined the mechanisms underlying primordial follicle activation in mice. We found that endothelial nitric oxide synthase (eNOS) and its downstream effectors, cyclic guanosine monophosphate (cGMP) and cGMP-dependent protein kinase G (PKG), were expressed in pre-granulosa cells and promoted primordial follicle activation, oocyte growth and granulosa cell proliferation in neonatal ovaries. Mammalian target of rapamycin (mTOR) colocalized with PKG in pre-granulosa cells and was essential for eNOS/cGMP/PKG pathway-induced primordial follicle activation. The eNOS/cGMP/PKG pathway was found to stabilize mTOR protein. The mRNA levels of F-box and WD repeat domain containing 7 (FBXW7), an E3 ubiquitin ligase, correlated negatively with mTOR protein levels in neonatal ovaries. FBXW7 bound to and destabilized mTOR protein in pre-granulosa cells in a ubiquitin/proteasome-dependent manner. However, agonists of the eNOS/cGMP/PKG pathway reduced FBXW7 mRNA levels. FBXW7 overexpression suppressed primordial follicle activation and prevented the eNOS/cGMP/PKG pathway from activating primordial follicles and stabilizing mTOR protein. These findings demonstrate that the eNOS/cGMP/PKG pathway activates primordial follicles by suppressing FBXW7-induced ubiquitination of mTOR in mice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Insulin-transferrin-sodium selenite media supplement, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
MDL-12,330A hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
8-Bromoguanosine 3′,5′-cyclic monophosphate sodium salt, ≥98% (HPLC), powder
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
YC-1, powder