Merck
  • Soil organic carbon, extracellular polymeric substances (EPS), and soil structural stability as affected by previous and current land-use.
Soil organic carbon, extracellular polymeric substances (EPS), and soil structural stability as affected by previous and current land-use.
Geoderma (2020-04-08)
M Redmile-Gordon, A S Gregory, R P White, C W Watts
ABSTRAKT
While soil microbial ecology, soil organic carbon (SOC) and soil physical quality are widely understood to be interrelated - the underlying drivers of emergent properties, from land management to biochemistry, are hotly debated. Biological binding agents, microbial exudates, or 'extracellular polymeric substances' (EPS) in soil are now receiving increased attention due to several of the existing methodological challenges having been overcome. We applied a recently developed approach to quantify soil EPS, as extracellular protein and extracellular polysaccharide, on the well-characterised soils of the Highfield Experiment, Rothamsted Research, UK. Our aim was to investigate the links between agricultural land use, SOC, transient binding agents known as EPS, and their impacts on soil physical quality (given by mean weight diameter of water stable aggregates; MWD). We compared the legacy effects from long-term previous land-uses (unfertilised grassland, fertilised arable, and fallow) which were established > 50 years prior to investigation, crossed with the same current land-uses established for a duration of only 2.5 years prior to sampling. Continuously fallow and grassland soils represented the poorest and greatest states of structural integrity, respectively. Total SOC and N were found to be affected by both previous and current land-uses, while extractable EPS and MWD were driven primarily by the current land-use. Land-use change between these two extremes (fallow → grass; grass → fallow) resulted in smaller SOC differences (64% increase or 37% loss) compared to MWD (125% increase or 78% loss). SOC concentration correlated well to MWD (adjusted R 2 = 0.72) but the greater SOC content from previous grassland was not found to contribute directly to the current stability (p < 0.05). Our work thus supports the view that certain distinct components of SOC, rather than the total pool, have disproportionately important effects on a soil's structural stability. EPS-protein was more closely related to aggregate stability than EPS-polysaccharide (p values of 0.002 and 0.027, respectively), and ranking soils with the 5 greatest concentrations of EPS-protein to their corresponding orders of stability (MWD) resulted in a perfect match. We confirmed that both EPS-protein and EPS-polysaccharide were transient fractions: supporting the founding models for aggregate formation. We suggest that management of transient binding agents such as EPS -as opposed to simply increasing the total SOC content- may be a more feasible strategy to improve soil structural integrity and help achieve environmental objectives.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Informacje o cenach i dostępności nie są obecnie dostępne.