Merck
Effects of Environmental Conditions on Nephron Number: Modeling Maternal Disease and Epigenetic Regulation in Renal Development.
International journal of molecular sciences (2021-05-01)
Lars Fuhrmann, Saskia Lindner, Alexander-Thomas Hauser, Clemens Höse, Oliver Kretz, Clemens D Cohen, Maja T Lindenmeyer, Wolfgang Sippl, Manfred Jung, Tobias B Huber, Nicola Wanner
ABSTRAKT
A growing body of evidence suggests that low nephron numbers at birth can increase the risk of chronic kidney disease or hypertension later in life. Environmental stressors, such as maternal malnutrition, medication and smoking, can influence renal size at birth. Using metanephric organ cultures to model single-variable environmental conditions, models of maternal disease were evaluated for patterns of developmental impairment. While hyperthermia had limited effects on renal development, fetal iron deficiency was associated with severe impairment of renal growth and nephrogenesis with an all-proximal phenotype. Culturing kidney explants under high glucose conditions led to cellular and transcriptomic changes resembling human diabetic nephropathy. Short-term high glucose culture conditions were sufficient for long-term alterations in DNA methylation-associated epigenetic memory. Finally, the role of epigenetic modifiers in renal development was tested using a small compound library. Among the selected epigenetic inhibitors, various compounds elicited an effect on renal growth, such as HDAC (entinostat, TH39), histone demethylase (deferasirox, deferoxamine) and histone methyltransferase (cyproheptadine) inhibitors. Thus, metanephric organ cultures provide a valuable system for studying metabolic conditions and a tool for screening for epigenetic modifiers in renal development.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-WT1, klon 6F-H2, clone 6F-H2, Upstate®, from mouse
Informacje o cenach i dostępności nie są obecnie dostępne.