Merck
Inhibition of phosphodiesterase type 9 reduces obesity and cardiometabolic syndrome in mice.
The Journal of clinical investigation (2021-10-08)
Sumita Mishra, Nandhini Sadagopan, Brittany Dunkerly-Eyring, Susana Rodriguez, Dylan C Sarver, Ryan P Ceddia, Sean A Murphy, Hildur Knutsdottir, Vivek P Jani, Deepthi Ashok, Christian U Oeing, Brian O'Rourke, Jon A Gangoiti, Dorothy D Sears, G William Wong, Sheila Collins, David A Kass
ABSTRAKT
Central obesity with cardiometabolic syndrome (CMS) is a major global contributor to human disease, and effective therapies are needed. Here, we show that cyclic GMP-selective phosphodiesterase 9A inhibition (PDE9-I) in both male and ovariectomized female mice suppresses preestablished severe diet-induced obesity/CMS with or without superimposed mild cardiac pressure load. PDE9-I reduces total body, inguinal, hepatic, and myocardial fat; stimulates mitochondrial activity in brown and white fat; and improves CMS, without significantly altering activity or food intake. PDE9 localized at mitochondria, and its inhibition in vitro stimulated lipolysis in a PPARα-dependent manner and increased mitochondrial respiration in both adipocytes and myocytes. PPARα upregulation was required to achieve the lipolytic, antiobesity, and metabolic effects of PDE9-I. All these PDE9-I-induced changes were not observed in obese/CMS nonovariectomized females, indicating a strong sexual dimorphism. We found that PPARα chromatin binding was reoriented away from fat metabolism-regulating genes when stimulated in the presence of coactivated estrogen receptor-α, and this may underlie the dimorphism. These findings have translational relevance given that PDE9-I is already being studied in humans for indications including heart failure, and efficacy against obesity/CMS would enhance its therapeutic utility.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
BAY 60-2770, ≥98% (HPLC)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Oleic acid, ≥99% (GC)