Merck
Antigenicity and Safety Evaluation of Lactiplantibacillus plantarum 7-2 Screened to Reduce α-Casein Antigen.
Foods (Basel, Switzerland) (2022-01-12)
Guangqing Mu, Zhao Zhang, Jiayi Wang, Shujuan Jiang, Hongxin Wang, Yunpeng Xu, Xinling Li, Lei Chi, Yue Li, Yanfeng Tuo, Xuemei Zhu
ABSTRAKT
α-Casein (α-CN) is considered the main allergen in bovine milk. Lactic acid bacteria (LAB) fermentation can hydrolyze milk protein and therefore reduce the antigenicity. In this paper, a LAB reducing the antigenicity of casein, identified as LactiplantibacillusPlantarum 7-2 (L. plantarum 7-2), was primarily identified by screening for protein hydrolysis ability using a method involving the determination of released free amino acid with further selection for the ideal antigenicity-reducing capability by enzyme-linked immunosorbent assay (ELISA). In order to verify the capability of L. plantarum 7-2 in inhibiting antigenicity, the standard milk proteins α-LA, β-LG, α-CN, β-CN and κ-CN were cultured with L. plantarum 7-2 for 18 h; The results of SDS-PAGE show that all the bands corresponding to the full length tested proteins became unclear or completely disappeared indicating that these proteins were hydrolyzed by L. plantarum 7-2. Correspondingly, the antigenicities of α-CN and β-LG were significantly reduced. L. plantarum 7-2 demonstrated negative hemolysis and nitrate reductase capabilities and was sensitive to the commonly used antibiotics ampicillin clindamycin tetracycline chloramphenicol, and erythromycin, demonstrating that L. plantarum 7-2 could be used in dairy product fermentation to reduce the antigenicity of milk protein.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Casein from bovine milk, BioUltra, ≥98% (PAGE)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
α-Lactalbumin from bovine milk, Type III, calcium depleted, ≥85% (PAGE), lyophilized powder
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
α-Casein from bovine milk, ≥70% αs-casein basis (electrophoresis), lyophilized powder
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥90% (PAGE), lyophilized powder
Informacje o cenach i dostępności nie są obecnie dostępne.