Merck
HK1 from hepatic stellate cell-derived extracellular vesicles promotes progression of hepatocellular carcinoma.
Nature metabolism (2022-10-04)
Qi-Tao Chen, Zhi-Yuan Zhang, Qiao-Ling Huang, Hang-Zi Chen, Wen-Bin Hong, Tianwei Lin, Wen-Xiu Zhao, Xiao-Min Wang, Cui-Yu Ju, Liu-Zheng Wu, Ya-Ying Huang, Pei-Pei Hou, Wei-Jia Wang, Dawang Zhou, Xianming Deng, Qiao Wu
ABSTRAKT
Extracellular vesicles play crucial roles in intercellular communication in the tumor microenvironment. Here we demonstrate that in hepatic fibrosis, TGF-β stimulates the palmitoylation of hexokinase 1 (HK1) in hepatic stellate cells (HSCs), which facilitates the secretion of HK1 via large extracellular vesicles in a TSG101-dependent manner. The large extracellular vesicle HK1 is hijacked by hepatocellular carcinoma (HCC) cells, leading to accelerated glycolysis and HCC progression. In HSCs, the nuclear receptor Nur77 transcriptionally activates the expression of depalmitoylase ABHD17B to inhibit HK1 palmitoylation, consequently attenuating HK1 release. However, TGF-β-activated Akt functionally represses Nur77 by inducing Nur77 phosphorylation and degradation. We identify the small molecule PDNPA that binds Nur77 to generate steric hindrance to block Akt targeting, thereby disrupting Akt-mediated Nur77 degradation and preserving Nur77 inhibition of HK1 release. Together, this study demonstrates an overlooked function of HK1 in HCC upon its release from HSCs and highlights PDNPA as a candidate compound for inhibiting HCC progression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
LX-2 Human Hepatic Stellate Cell Line, a highly suitable model of human hepatic fibrosis
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
Gö 6983, ≥97%
Roche
Anty-Digoxigenin-AP, fragmenty Fab, from sheep