Merck
  • Bioaccessibility assay, antioxidant activity and consumer-oriented sensory analysis of Beta vulgaris by-product encapsulated in Ca(II)-alginate beads for different foods.
Bioaccessibility assay, antioxidant activity and consumer-oriented sensory analysis of Beta vulgaris by-product encapsulated in Ca(II)-alginate beads for different foods.
Food chemistry. Molecular sciences (2022-10-25)
Tatiana Rocio Aguirre-Calvo, Natalia Sosa, Tamara Anahí López, María Ximena Quintanilla-Carvajal, Mercedes Perullini, Patricio Román Santagapita
ABSTRAKT
Bioaccessibility analysis and antioxidant activity along in vitro digestion and a consumer-oriented sensory analysis were conducted in three potential functional foods based on Ca(II)-alginate beads containing bioactive compounds extracted from beet stems. Ca(II)-alginate beads per se, and two selected products (cookies and turkish delights supplemented with the beads) were prepared. Regarding the beads, among the attributes rated by consumers, visual appreciation predominates, being color in the just-as-right (JAR) category and in the like preference. Instead, both flavor and sweet taste were attributes highly penalized and should be improved in beads to be accepted as food per se. A higher percentage of customers preferred cookies and turkish delights instead of only beads, considering global satisfaction. Regarding in vitro digestion, there was a significant content of phenolic compounds in the products with beads, showing a bioaccessibility greater than 80% (for cookies) and 26% (for turkish delights). Also, the antioxidant capacity measured by ABTS ranged between 50 and 109% for cookies and turkish delights, being lower when measured by FRAP (between 20 and 30%, respectively). Thus, including the beads with beet stem extract in both products leads to a significant increase in the content of phenolic compounds and in the antioxidant capacity compared to their counterparts, protecting the compound during oral and gastric phases. These results allow the generation of improved Ca(II)-alginate systems with promising functional properties for the development of ingredients and functional foods.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Informacje o cenach i dostępności nie są obecnie dostępne.