Merck
PD-1 instructs a tumor-suppressive metabolic program that restricts glycolysis and restrains AP-1 activity in T cell lymphoma.
Nature cancer (2023-09-19)
Tim Wartewig, Jay Daniels, Miriam Schulz, Erik Hameister, Abhinav Joshi, Joonhee Park, Emma Morrish, Anuroop V Venkatasubramani, Filippo M Cernilogar, Frits H A van Heijster, Christian Hundshammer, Heike Schneider, Filippos Konstantinidis, Judith V Gabler, Christine Klement, Henry Kurniawan, Calvin Law, Yujin Lee, Sara Choi, Joan Guitart, Ignasi Forne, Jérôme Giustinani, Markus Müschen, Salvia Jain, David M Weinstock, Roland Rad, Nicolas Ortonne, Franz Schilling, Gunnar Schotta, Axel Imhof, Dirk Brenner, Jaehyuk Choi, Jürgen Ruland
ABSTRAKT
The PDCD1-encoded immune checkpoint receptor PD-1 is a key tumor suppressor in T cells that is recurrently inactivated in T cell non-Hodgkin lymphomas (T-NHLs). The highest frequencies of PDCD1 deletions are detected in advanced disease, predicting inferior prognosis. However, the tumor-suppressive mechanisms of PD-1 signaling remain unknown. Here, using tractable mouse models for T-NHL and primary patient samples, we demonstrate that PD-1 signaling suppresses T cell malignancy by restricting glycolytic energy and acetyl coenzyme A (CoA) production. In addition, PD-1 inactivation enforces ATP citrate lyase (ACLY) activity, which generates extramitochondrial acetyl-CoA for histone acetylation to enable hyperactivity of activating protein 1 (AP-1) transcription factors. Conversely, pharmacological ACLY inhibition impedes aberrant AP-1 signaling in PD-1-deficient T-NHLs and is toxic to these cancers. Our data uncover genotype-specific vulnerabilities in PDCD1-mutated T-NHL and identify PD-1 as regulator of AP-1 activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
BMS-303141, ≥98% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Przeciwciało anty-acetylo-histonowe H4, 1 mg/mL, Upstate®