Merck
2D-PAGE as an effective method of RNA degradome analysis.
Molecular biology reports (2011-05-12)
Martyna Nowacka, Paulina Jackowiak, Agnieszka Rybarczyk, Tomasz Magacz, Pawel M Strozycki, Jan Barciszewski, Marek Figlerowicz
ABSTRAKT
The continuously growing interest in small regulatory RNA exploration is one of the important factors that have inspired the recent development of new high throughput techniques such as DNA microarrays or next generation sequencing. Each of these methods offers some significant advantages but at the same time each of them is expensive, laborious and challenging especially in terms of data analysis. Therefore, there is still a need to develop new analytical methods enabling the fast, simple and cost-effective examination of the complex RNA mixtures. Recently, increasing attention has been focused on the RNA degradome as a potential source of riboregulators. Accordingly, we attempted to employ a two-dimensional gel electrophoresis as a quick and uncomplicated method of profiling RNA degradome in plant or human cells. This technique has been successfully used in proteome analysis. However, its application in nucleic acids studies has been very limited. Here we demonstrate that two dimensional electrophoresis is a technique which allows one to quickly and cost-effectively identify and compare the profiles of 10-90 nucleotide long RNA accumulation in various cells and organs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture