- Lipoprotein lipase in chronic lymphocytic leukaemia - strong biomarker with lack of functional significance.
Lipoprotein lipase in chronic lymphocytic leukaemia - strong biomarker with lack of functional significance.
Leukemia research (2013-03-13)
Edit Porpaczy, Stefanie Tauber, Martin Bilban, Gerhard Kostner, Michaela Gruber, Sandra Eder, Daniel Heintel, Trang Le, Karin Fleiss, Cathrin Skrabs, Medhat Shehata, Ulrich Jäger, Katrina Vanura
PMID23478142
ABSTRAKT
In chronic lymphocytic leukaemia (CLL), lipoprotein lipase (LPL) mRNA overexpression is an established poor prognostic marker, its function, however, is poorly understood. Measuring extracellular LPL enzymatic activity and protein, we found no difference between levels in CLL patients and those of controls, both before and after heparin treatment in vivo and in vitro. Investigating LPL knock down effects, we determined five potential downstream targets, of which one gene, STXBP3, reportedly is involved in fatty acid metabolism. While possibly reflecting an epigenetic switch towards an incorrect transcriptional program, LPL overexpression by itself does not appear to significantly influence CLL cell survival.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lipoprotein Lipase from bovine milk, ammonium sulfate suspension, ≥2,000 units/mg protein (BCA)
Sigma-Aldrich
Lipoprotein Lipase from Pseudomonas sp., lyophilized, powder, yellow-brown, ≥160 U/mg
Sigma-Aldrich
Lipoprotein Lipase from Burkholderia sp., lyophilized powder, ≥50,000 units/mg solid