Merck
Analysis of a genetic mutation in an electrophoretic variant of the human lactate dehydrogenase-B(H) subunit.
Human genetics (1993-06-01)
M Maekawa, K Sudo, M Kitajima, Y Matsuura, S S Li, T Kanno
ABSTRAKT
An electrophoretic variant of the lactate dehydrogenase (LDH)-B(H) subunit was discovered in a patient with diabetes mellitus. His LDH activity in serum was slightly lower than normal and the LDH isozyme pattern showed an abnormal migration indicating an LDH-B subunit variant of the fast type. The LDH containing the variant subunit revealed a decreased heat stability. DNA analysis of the variant allele detected a base substitution, an A to G transition, at codon 6 (AAA-->GAA). The mutation resulted in the replacement of a lysine by a glutamic acid (K6E). The change may cause the heat instability and affect the net charge of the variant subunit, resulting in an electrophoretic LDH-B subunit variant of the fast type.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein