Merck
Metal sulfide nanoparticles synthesized via enzyme treatment of biopolymer stabilized nanosuspensions.
Nanoscale (2010-07-21)
Yi-Yeoun Kim, Dominic Walsh
ABSTRAKT
Nanoparticles of CuS, Cu(x)S, Ag(2)S and CdS were successfully prepared using a novel general and green synthetic process to give dextran biopolymer stabilised metal sulfifde nanosuspensions. Following preparation, dextranase enzyme was used to remove the bulk of the bound dextran to give pure stable metal sulfide nanocrystals for application in for example aspects of medicine, photonics and solar cells. Particles of good homogeneity were obtained and the CuS nanoparticle size was controlled to 9-27 nm by adjusting the reaction conditions. Cu(2)S nanoparticles were 14 nm, Ag(2)S nanoparticles were 20-50 nm and CdS nanoparticles were 9 nm is size. The complexing mechanism of nanoparticle sulfides to dextrans was further studied using carboxylmethyl dextran as a complexing agent and crosslinked Sephadex (dextran) ;beads as substrate. Particles were characterized by TEM, XRD, TGA, FT-IR and zeta-potential measurement, and their UV-vis spectroscopic absorption properties were determined. Stabilization of the sulfide nanoparticles with soluble hydroxylated biopolymers such as dextran is previously unreported and is here interpreted in terms of viscosity, pH of the system and weak polar S-H or S(metal)OH(2)(+) interactions with dextran depending on the material. Notably, the complexing mechanism appears to differ significantly from that taking place in known dextran-metal oxide systems. The process shown here has good potential for scale-up as a biosynthetic route for a range of functional sulfide nanoparticles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Copper(II) sulfide, powder, 99.99% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Copper(II) sulfide, powder, −100 mesh, ≥99% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.