Merck
  • Vitamins K interact with N-terminus α-synuclein and modulate the protein fibrillization in vitro. Exploring the interaction between quinones and α-synuclein.
Vitamins K interact with N-terminus α-synuclein and modulate the protein fibrillization in vitro. Exploring the interaction between quinones and α-synuclein.
Neurochemistry international (2012-10-16)
Fernanda Luna da Silva, Eduardo Coelho Cerqueira, Mônica Santos de Freitas, Daniela Leão Gonçalves, Lilian Terezinha Costa, Cristian Follmer
ABSTRAKT
In the last decades, a series of compounds, including quinones and polyphenols, has been described as having anti-fibrillogenic action on α-synuclein (α-syn) whose aggregation is associated to the pathogenesis of Parkinson's disease (PD). Most of these molecules act as promiscuous anti-amyloidogenic agents, interacting with the diverse amyloidogenic proteins (mostly unfolded) through non-specific hydrophobic interactions. Herein we investigated the effect of the vitamins K (phylloquinone, menaquinone and menadione), which are 1,4-naphthoquinone (1,4-NQ) derivatives, on α-syn aggregation, comparing them with other anti-fibrillogenic molecules such as quinones, polyphenols and lipophilic vitamins. Vitamins K delayed α-syn fibrillization in substoichiometric concentrations, leading to the formation of short, sheared fibrils and amorphous aggregates, which are less prone to produce leakage of synthetic vesicles. In seeding conditions, menadione and 1,4-NQ significantly inhibited fibrils elongation, which could be explained by their ability to destabilize preformed fibrils of α-syn. Bidimensional NMR experiments indicate that a specific site at the N-terminal α-syn (Gly31/Lys32) is involved in the interaction with vitamins K, which is corroborated by previous studies suggesting that Lys is a key residue in the interaction with quinones. Together, our data suggest that 1,4-NQ, recently showed up by our group as a potential scaffold for designing new monoamine oxidase inhibitors, is also capable to modulate α-syn fibrillization in vitro.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Phytonadione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Sigma-Aldrich
1,4-Naphthoquinone, 97%
Sigma-Aldrich
1,4-Naphthoquinone, purum, ≥96.5% (HPLC)
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Vitamin K1, viscous liquid
Supelco
Menadione (K3), analytical standard
Phytomenadione, European Pharmacopoeia (EP) Reference Standard
Menadione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Menadione sodium bisulfite, BioReagent, suitable for cell culture, ≥95% (TLC)
Sigma-Aldrich
Menadione sodium bisulfite, ≥95% (TLC)
Supelco
Phylloquinone (K1), analytical standard