Merck
Identification of cell wall proteins in the flax (Linum usitatissimum) stem.
Proteomics (2013-01-03)
Arnaud Day, Stéphane Fénart, Godfrey Neutelings, Simon Hawkins, Christian Rolando, Caroline Tokarski
ABSTRAKT
Sequential salt (CaCl2 , LiCl) extractions were used to obtain fractions enriched in cell wall proteins (CWPs) from the stem of 60-day-old flax (Linum usitatissimum) plants. High-resolution FT-ICR MS analysis and the use of recently published genomic data allowed the identification of 11 912 peptides corresponding to a total of 1418 different proteins. Subcellular localization using TargetP, Predotar, and WoLF PSORT led to the identification of 152 putative flax CWPs that were classified into nine different functional classes previously established for Arabidopsis thaliana. Examination of different functional classes revealed the presence of a number of proteins known to be involved in, or potentially involved in cell-wall metabolism in plants. The flax stem cell wall proteome was also compared with transcriptomic data previously obtained on comparable samples. This study represents a major contribution to the identification of CWPs in flax and will lead to a better understanding of cell wall biology in this species.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, Molecular Biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis