Merck
Cell death in the skin: how to study its quality and quantity?
Methods in molecular biology (Clifton, N.J.) (2013-01-18)
Roman Makarov, Peter Geserick, Maria Feoktistova, Martin Leverkus
ABSTRAKT
The characterization of the quality and quantity of cell death has gained substantial interest over the past decades. More recently necroptosis as a programmed form of necrosis has been identified as an important additional form of cell death with relevance in the skin. Understanding how to assay cell death in specific is of critical importance for cancer research and treatment. Here we describe six different methods that can be used to assay cell viability and to study the quality or quantity of cultured human keratinocytes in vitro. These methods include crystal violet assay, hypodiploidy analysis, caspase-8 cleavage, release of HMGB1, annexin V/propidium iodide co-staining, and Hoechst/SYTOX green co-staining.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Crystal Violet, suitable for microscopy
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, S. No.: 785
Sigma-Aldrich
Crystal Violet Solution
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution