Merck
Preparation and characterization of anti-algal sustained-release granules and their inhibitory effects on algae.
Chemosphere (2013-01-29)
Lixiao Ni, Kumud Acharya, Gaoxiang Ren, Shiyin Li, Yiping Li, Yong Li
ABSTRAKT
The objectives of this work were to prepare and characterize an anti-algal sustained-release granule, then study its mode of action on Microcystis aeruginosa. The anti-algal sustained-release granule was prepared with artemisinin using alginate-chitosan microcapsule technology and characterized by a high performance liquid chromatography with an evaporative light-scattering detector, Fourier transform infrared spectral analysis, and a scanning electron microscope. The optimum preparation (in %, w/v) using the orthogonal method was: 2.5 sodium alginate; 0.25 chloride; 0.6 artemisinin; 2 calcium chloride; and 1.5 mL of the cross-linking agent, glutaraldehyde. These artemisinin sustained-release granules had a high encapsulation efficiency (up to 68%) and good release properties (release time of more than 40 d). Artemisinin sustained-release granules released cumulatively in a solution containing M. aeruginosa, and the stress on algae increased gradually within 30 d. Artemisinin sustained-release granules decreased the content of the soluble protein, Chlorophyll a in 30 d, increased the superoxide dismutase activity of M. aeruginosa, but exerted no effect on the soluble sugar content. Compared to direct dosing of artemisinin, algae can be inhibited longer and more effectively by the artemisinin sustained-release granules. The results of our research can aid in the development of new anti-algal sustained-release granules and lead to further study of their application in the field.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, Molecular Biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Artemisinin, 98%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis