Merck
  • Enantioselective synthesis of multisubstituted biaryl skeleton by chiral phosphoric acid catalyzed desymmetrization/kinetic resolution sequence.
Enantioselective synthesis of multisubstituted biaryl skeleton by chiral phosphoric acid catalyzed desymmetrization/kinetic resolution sequence.
Journal of the American Chemical Society (2013-02-19)
Keiji Mori, Yuki Ichikawa, Manato Kobayashi, Yukihiro Shibata, Masahiro Yamanaka, Takahiko Akiyama
ABSTRAKT
Described herein is the enantioselective synthesis of multisubstituted biaryl derivatives by chiral phosphoric acid catalyzed asymmetric bromination. Two asymmetric reactions (desymmetrization and kinetic resolution) proceeded successively to afford chiral biaryls in excellent enantioselectivities (up to 99% ee). Both experimental and computational studies suggested that this excellent selectivity could be achieved via a highly organized hydrogen bond network among a substrate, a catalyst (chiral phosphoric acid), and a brominating reagent (N-bromophthalimide).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
N-Bromophthalimide, 95%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis