Merck
C-reactive protein and fibrin clot strength measured by thrombelastography after coronary stenting.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis (2013-02-23)
Rolf P Kreutz, Janelle Owens, Jeffrey A Breall, Deshun Lu, Elisabeth von der Lohe, Islam Bolad, Anjan Sinha, David A Flockhart
ABSTRAKT
Inflammation is implicated in the progression of coronary artery disease and the molecular processes of inflammation and thrombosis are closely intertwined. Elevated levels of C-reactive protein (CRP) have been associated with an elevated risk of adverse ischaemic events after coronary stenting and hypercoagulability. Heightened whole blood clot strength measured by thrombelastography (TEG) has been associated with adverse ischaemic events after stenting. We intended to examine the relationship of CRP to plasma fibrin clot strength in patients after coronary stenting. Plasma fibrin clot strength was measured by TEG in 54 patients 16-24 h after undergoing elective percutaneous coronary intervention (PCI). Coagulation was induced in citrated plasma by addition of kaolin and CaCl2. Plasma levels of CRP and fibrinogen were measured by enzyme-linked immunoassay. Increasing quartiles of CRP were associated with increasing levels of maximal plasma fibrin clot strength measured by TEG (P < 0.001) and increasing BMI (P = 0.04). Patients in the highest quartile of CRP had significantly higher maximal fibrin clot strength (G) than the patients in the lowest quartile (G: 3438 ± 623 vs. 2184 ± 576 dyn/cm, P < 0.0001). Fibrinogen concentration was not significantly different across quartiles of CRP (P = 0.97). Patients with established coronary artery disease undergoing coronary stenting who have elevated CRP after PCI exhibit heightened maximal plasma fibrin clot strength as compared with those with low CRP. Thrombotic risk associated with elevated CRP may be linked to procoagulant changes and high tensile fibrin clot strength independent of fibrinogen concentration.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, Molecular Biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Kaolin, meets USP testing specifications
Sigma-Aldrich
Kaolin, heavy, powder
Sigma-Aldrich
Halloysite nanoclay
Sigma-Aldrich
Kaolin, tested according to Ph. Eur.
Sigma-Aldrich
Kaolin
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis