Merck
Propylthiouracil-induced hypothyroidism during lactation alters leucine and mannose metabolism in rat cerebellar slices.
Experimental biology and medicine (Maywood, N.J.) (2013-03-13)
Alexandre Pastoris Müller, Aline Longoni, Marcelo Farina, Clarice K B da Silveira, Diogo Onofre Souza, Marcos Luiz Santos Perry, Adriano Martimbianco de Assis
ABSTRAKT
Thyroid hormone deficiency during perinatal development results in significant alterations in neurological functions. The relationship between such events and brain metabolism is not completely understood. The aim of this study was to investigate the effects of hypothyroidism on leucine, mannose, glucose and lactate metabolism in rat cerebellar slices. Experimental hypothyroidism was induced by exposing mothers and pups to propylthiouracil (PTU) until weaning - postnatal day 21. Metabolic analyses were performed in postnatal day 10 (PND10) and 21 (PND21) animals. A matching group of animals received the same oral treatment also after weaning until adulthood PND60 with T3 supplement during lactation (P1-P21). In PND21 animals, PTU treatment significantly increased the rate of leucine oxidation to CO2, although glucose and lactate oxidations were not affected. PTU treatment also increased the oxidation of leucine to CO2 at PND60 (adult animals). PND10 hypothyroidism animals showed a decrease in conversion of mannose to glycolipids and glycoprotein compared with the control group. However, PTU treatment increased the conversion of mannose to glycolipids and glycoprotein in PND21 animals. The replacement of T3 normalized mannose and leucine metabolism in adult rats. These results indicate that deficits in thyroid hormones during lactation could delay or alter brain development and metabolism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6-Propyl-2-thiouracil, enzyme inhibitor
Supelco
6-Propyl-2-thiouracil, VETRANAL®, analytical standard
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, 99%, FG
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland