Merck
  • Mutations in adenosine deaminase-like (ADAL) protein confer resistance to the antiproliferative agents N6-cyclopropyl-PMEDAP and GS-9219.
Mutations in adenosine deaminase-like (ADAL) protein confer resistance to the antiproliferative agents N6-cyclopropyl-PMEDAP and GS-9219.
Anticancer research (2013-05-07)
Christian R Frey, Graciela Andrei, Ivan Votruba, Carina Cannizzaro, Bin Han, Wanchi Fung, Magdeleine Hung, Xiaohong Liu, Romas Geleziunas, Pierre Fiten, Ghislain Opdenakker, Robert Snoeck, Tomas Cihlar
ABSTRAKT
GS 9219 is a double prodrug of antiproliferative nucleotide analog 9-(2-Phosphonylmethoxyethyl)guanine (PMEG), with potent in vivo efficacy against various hematological malignancies. This study investigates the role of adenosine deaminase-like (ADAL) protein in the intracellular activation of GS-9219. A cell line resistant to 9-(2-Phosphonylmethoxyethyl)-N(6)-cyclopropyl-2,6-diaminopurine (cPrPMEDAP), an intermediate metabolite of GS-9219, was generated and characterized. The resistant cell line was cross-resistant to cPrPMEDAP and GS-9219, due to a defect in the deamination of cPrPMEDAP to PMEG. Mutations in the ADAL gene (H286R and S180N) were identified in the resistant cells that adversely-affected its enzymatic activity. Introduction of the wild-type ADAL gene re-sensitized resistant cells to both cPrPMEDAP and GS-9219. The ADAL protein plays an essential role in the intracellular activation of GS-9219 by catalyzing the deamination of cPrPMEDAP metabolite to PMEG. Mutations affecting the activity of ADAL confer resistance to both GS-9219 and its metabolite cPrPMEDAP.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Alanine, ≥99%
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C