Merck
Update to Interim Guidance for Preexposure Prophylaxis (PrEP) for the Prevention of HIV Infection: PrEP for injecting drug users.
MMWR. Morbidity and mortality weekly report (2013-06-14)
ABSTRAKT
On June 12, 2013, the Thailand Ministry of Health and CDC published results from a randomized controlled trial of a daily oral dose of 300 mg of tenofovir disoproxil fumarate (TDF) that showed efficacy in reducing the acquisition of human immunodeficiency virus (HIV) infection among injecting drug users (IDUs) (1). Based on these findings, CDC recommends that preexposure prophylaxis (PrEP) be considered as one of several prevention options for persons at very high risk for HIV acquisition through the injection of illicit drugs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Adenine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenine, ≥99%