Merck
  • Identification and characterization of two peptidoglycan recognition proteins with zinc-dependent antibacterial activity from the cotton bollworm, Helicoverpa armigera.
Identification and characterization of two peptidoglycan recognition proteins with zinc-dependent antibacterial activity from the cotton bollworm, Helicoverpa armigera.
Developmental and comparative immunology (2013-01-09)
Dai-Qun Yang, Zhen-Lin Su, Chuan Qiao, Zan Zhang, Jia-Lin Wang, Fei Li, Xu-Sheng Liu
ABSTRAKT
Peptidoglycan recognition proteins (PGRPs) specifically bind to peptidoglycan and play an important role in the innate immune responses as pattern recognition receptors (PRRs). Here we identified and characterized two PGRPs (HaPGRP-B and HaPGRP-C) from the cotton bollworm, Helicoverpa armigera. The comparative analysis indicated that five amino acids which are required for T7 lysozyme Zn(2+) binding and amidase activity are conserved in HaPGRP-B and HaPGRP-C, suggesting that the two PGRPs are members of the amidase-type PGRPs. HaPGRP-B and HaPGRP-C mRNA increased in both the fat bodies and the hemocytes after an injection of Gram-negative Escherichia coli or Gram-positive Staphylococcus aureus. Recombinant HaPGRP-B and HaPGRP-C could agglutinate E. coli and S. aureus in a zinc-dependent manner. More importantly, both rHaPGRP-B and rHaPGRP strongly inhibited the growth of E. coli and S. aureus in the presence of Zn(2+). Moreover, the HaPGRP-B mRNA showed up-regulation post hormones (20E and methoprene) injection. Our results indicate that the two PGRPs of H. armigera may play an important role in defending against bacteria as amidase-type PGRPs and the hormones can function in regulating the expressions of PGRPs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Amidase from Pseudomonas aeruginosa, recombinant, expressed in E. coli, buffered aqueous glycerol solution, hydroxamate transferase ≥200 units/mg protein (biuret)
Informacje o cenach i dostępności nie są obecnie dostępne.