Merck
A smart approach to add antibacterial functionality to cellulosic pigment prints.
Carbohydrate polymers (2013-04-03)
N A Ibrahim, B M Eid, T M Abou Elmaaty, E Abd El-Aziz
ABSTRAKT
This study was devoted to enhancing the antibacterial functionality of pigment printed cotton, linen and viscose fabrics. Ag-NP's/PVP colloid, triclosan derivatives, chitosan or choline chloride was successfully incorporated into the pigment paste followed by printing and microwave curing to impart antibacterial activity to the cellulosic prints. Results obtained demonstrate that the modified pigment prints exhibit a remarkable antibacterial activity against the G+ve (Staphylococcus aureus) and G-ve (Escherichia coli) bacteria with a noticeable durability after 20 washing cycles without adversely affecting the printing and softness properties. The extent of printability and functionality of the nominated substrates are significantly governed by the type of: bio-active ingredient, binder, pigment and substrate. TEM, SEM and EDX analysis confirmed the formation of Ag-NP's/PVP colloid, of particle size range 7-14 nm, deposition of cross-linked-binder film onto the modified pigment prints, and the existence of elementary Ag and Si loaded onto fabrics surface, respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material