Merck
Halobellus inordinatus sp. nov., from a marine solar saltern and an inland salt lake of China.
International journal of systematic and evolutionary microbiology (2013-06-04)
Xing-Xing Qiu, Yun-Zhuang Mou, Mei-Lin Zhao, Wen-Jiao Zhang, Dong Han, Min Ren, Heng-Lin Cui
ABSTRAKT
Two halophilic archaeal strains, YC20(T) and XD15, were isolated from a marine solar saltern and an inland salt lake in China. Both had pleomorphic cells that lysed in distilled water, stained Gram-negative and formed red-pigmented colonies. They were neutrophilic, requiring at least 100 g NaCl l(-1) and 0.5-95 g MgCl2 l(-1) for growth at the optimum growth temperature of 37 °C. The major polar lipids of the two strains were phosphatidylglycerol (PG), phosphatidylglycerol phosphate methyl ester (PGP-Me), phosphatidylglycerol sulfate (PGS) and two major glycolipids chromatographically identical to sulfated mannosyl glucosyl diether (S-DGD-1) and mannosyl glucosyl diether (DGD-1), respectively. Trace amounts of two unidentified glycolipids were also detected. The 16S rRNA gene sequences of the two strains were 99.5 % identical and showed 94.0-95.9 % similarity to the most closely related members of the genus Halobellus of the family Halobacteriaceae. The rpoB' gene sequence similarity between strains YC20(T) and XD15 was 98.2 % and these sequences showed 89.6-92.8 % similarity to those of the most closely related members of the genus Halobellus. The DNA G+C contents of strains YC20(T) and XD15 were 65.8 mol% and 65.4 mol%, respectively. The DNA-DNA hybridization value between strain YC20(T) and strain XD15 was 92 %, and the two strains showed low DNA-DNA relatedness to members of the genus Halobellus. The phenotypic, chemotaxonomic and phylogenetic properties suggest that strains YC20(T) and XD15 represent a novel species of the genus Halobellus, for which the name Halobellus inordinatus sp. nov. is proposed. The type strain is YC20(T) ( = CGMCC 1.12120(T) = JCM 18361(T)) and the other strain is XD15 ( = CGMCC 1.12236 = JCM 18648).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis