Merck
Combined effects of potassium sorbate, hot water and thiabendazole against green mould of citrus fruit and residue levels.
Food chemistry (2013-06-26)
Salvatore D'Aquino, Angela Fadda, Antonio Barberis, Amedeo Palma, Alberto Angioni, Mario Schirra
ABSTRAKT
Postharvest treatments of potassium sorbate only controlled recently established infections of Penicillium digitatum on Femminello siracusano lemons but did not confer any persistent protection. The loss of efficacy of potassium sorbate to control green mould decay was related to its irregular deposition on the fruit surface, as revealed by environmental scanning electron microscopy of oranges, and to the brief persistence of potassium sorbate residues. When treatment was done at 53°C, the co-application of potassium sorbate with thiabendazole reduced thiabendazole residues in Moro and Sanguinello oranges, compared to thiabendazole treatment alone. However, treatment efficacy against two isolates of P. digitatum (thiabendazole-sensitive and thiabendazole-resistant) notably improved, indicating that potassium sorbate and hot water potentiated thiabendazole activity. Potassium sorbate residues remarkably decreased during fruit storage and were not affected by the co-application of thiabendazole.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Thiabendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thiabendazole, BioReagent, suitable for plant cell culture, powder
Supelco
Thiabendazole, PESTANAL®, analytical standard
Sigma-Aldrich
Thiabendazole, ≥99%, powder
Tiabendazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sorbic acid, 99.0-101.0% anhydrous basis
Sorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sorbic acid, tested according to Ph. Eur.