Merck
Assessment of serum S100B and neuron specific enolase levels to evaluate the neurotoxıc effects of organıc solvent exposure.
Clinical toxicology (Philadelphia, Pa.) (2013-07-31)
Cevdet Züngün, Fatma Meriã Yilmaz, Engіn Tutkun, Hinã Yilmaz, Sema Uysal
ABSTRAKT
Long-term organic solvent exposure may cause toxic effects in central nervous system . Trichloroethylene (TCE) is known to be one of the neurotoxic chlorinated organic solvents. Trichloroacetic acid (TCA) is an oxidative pathway metabolite of TCE. S100B, a calcium-binding protein in glial cells, and neuron specific enolase (NSE) in neuron cytoplasma are protein markers of astrocyte and neuron damage, respectively. Clinical and laboratory assesments were performed in 25 participants with organic solvent exposure history. Control group included 25 healthy age and sex-matched individuals. Measurements of serum S100B and NSE were performed using Roche Cobas E 601 compatible kits and elechtrochemiluminescence immunoassay. The levels of TCA in urine were measured by the headspace GC technique, after methyl esterification by methanol. Median value of urine TCA in solvent-exposed group was 12.30 mg/L with 10.20 mg/L and 35.00 mg/L minimum and maximum values, respectively. The difference between serum S100B levels of solvent-exposed group (0.064 μg/L) and control group (0.049 μg/L) was statistically significant (p < 0.05). Serum NSE levels of control group (15.61 ng/ml) were higher than solvent-exposed group (13.90 ng/ml) but difference was not statistically significant (p > 0.05). Serum S100B levels were found to be higher in solvent-exposed group when compared with control group. NSE levels were comparable between two groups. Increased Serum S100B levels in organic solvent exposure may indicate a preventive response to neuronal damage caused by reactive oxygen species (ROS) produced through oxidative metabolic pathways of organic solvents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Trichloroethylene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Trichloroethylene, ACS reagent, ≥99.5%
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Supelco
Trichloroethylene, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N