Merck
Cordycepin: a bioactive metabolite with therapeutic potential.
Life sciences (2013-10-15)
Hardeep S Tuli, Anil K Sharma, Sardul S Sandhu, Dharambir Kashyap
ABSTRAKT
Cytotoxic nucleoside analogues were the first chemotherapeutic agents for cancer treatment. Cordycepin, an active ingredient of the insect fungus Cordyceps militaris, is a category of compounds that exhibit significant therapeutic potential. Cordycepin has many intracellular targets, including nucleic acid (DNA/RNA), apoptosis and cell cycle, etc. Investigations of the mechanism of anti-cancer drugs have yielded important information for the design of novel drug targets in order to enhance anti-tumor activity with less toxicity to patients. This extensive review covers various molecular aspects of cordycepin interactions with its recognized cellular targets and proposes the development of novel therapeutic strategies for cancer treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cordycepin, from Cordyceps militaris
Informacje o cenach i dostępności nie są obecnie dostępne.