Merck
Mucosal protective agents prevent exacerbation of NSAID-induced small intestinal lesions caused by antisecretory drugs in rats.
The Journal of pharmacology and experimental therapeutics (2013-11-21)
Hiroshi Satoh, Kikuko Amagase, Koji Takeuchi
ABSTRAKT
Antisecretory drugs such as histamine H₂-receptor antagonists and proton pump inhibitors are commonly used for the treatment of upper gastrointestinal mucosal lesions induced by nonsteroidal anti-inflammatory drugs (NSAIDs). However, it has recently been reported that these drugs exacerbate NSAID-induced small intestinal lesions in rats. Unfortunately, there are few effective agents for the treatment of this complication. We examined the effects of mucosal protective agents (MPAs) (misoprostol, irsogladine, and rebamipide) and mucin of porcine stomach on diclofenac-induced intestinal lesions and the exacerbation of the lesions by ranitidine or omeprazole. The effects of the drugs on intestinal motility and mucus distribution/content were also examined. Male Wistar rats (180-220 g) were used. Each drug was administered orally under fed conditions. Diclofenac (1-10 mg/kg) produced multiple lesions in the small intestine dose-dependently. Both ranitidine (30 mg/kg) and omeprazole (100 mg/kg) significantly increased the intestinal lesions induced by low doses (3 and 6 mg/kg) of diclofenac. Misoprostol (0.03-0.3 mg/kg), irsogladine (3-30 mg/kg), and rebamipide (30-300 mg/kg), as well as mucin (30-300 mg/kg) inhibited the formation of intestinal lesions caused by a high dose (10 mg/kg) of diclofenac alone and prevented the exacerbation of diclofenac-induced lesions by antisecretory drugs. Diclofenac (10 mg/kg) markedly increased the intestinal motility and decreased the mucosal mucus, and the decrease of mucus was significantly inhibited by the MPAs. These results indicate the usefulness of the MPAs for the treatment of intestinal lesions induced by NSAIDs alone or by coadministration with antisecretory drugs, and suggest that mucus plays an important role in the protection of intestinal mucosa by the MPAs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Omeprazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Ranitidine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
Diclofenac sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Omeprazole, solid
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Omeprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ranitidine hydrochloride, solid
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Misoprostol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Misoprostol, ≥99% (HPLC)
Sigma-Aldrich
Diclofenac sodium salt
Supelco
Diclofenac sodium salt, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Alanine, ≥99%
Alanine, European Pharmacopoeia (EP) Reference Standard
Misoprostol for system suitability, European Pharmacopoeia (EP) Reference Standard