Merck
In vitro anti cancer activity of ethanol extract fractions of Aerva lanata L.
Pakistan journal of biological sciences : PJBS (2014-02-12)
Abhishek Bhanot, Rohini Sharma, Satnam Singh, Malleshappa N Noolvi, Shashank Singh
ABSTRAKT
To explore in vitro anticancer potential of Aerva lanata L. (flowering aerial part). The study was performed with 5 different human cell lines for the study of lung, leukaemia, prostate, colon and cervix cancer by using Sulphorhodamine B (SRB) assay. There were three doses of 10, 30 and 100 microg mL(-1) of each Aerva lanata L. Chloroform fraction (ALCF) and Aerva lanata L. Ethyl Acetate Fraction (ALEAF) used in this study. ALCF showed significant % inhibitory effect for leukaemia, lung and colon cancer at maximum concentration of 100 microg mL(-1) as compared to standard drug mitomycin. On the other hand ALEAF showed the significant % inhibitory effect for lung and cervix cancer at maximum concentration of 100 microg mL(-1) as compared to standard drug 5-fluoro Uracil (5-FU). From the above studies it is concluded that, the ethyl acetate fraction and chloroform fraction of Aerva lanata L. provide enough experimental evidence for anticancer activity and these fractions could be useful in medical care.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%