Merck
The effect of social class on the prevalence of caries, plaque, gingivitis and pocketing in 11-12-year-old children in South Wales.
Journal of dentistry (1987-10-01)
P M Dummer, M Addy, R Hicks, A Kingdon, W C Shaw
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Lactoglobulin A from bovine milk, Isoelectric focusing marker, pI 5.1
Sigma-Aldrich
β-Lactoglobulin A from bovine milk, ≥90% (PAGE)
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥90% (PAGE), lyophilized powder
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥90% (PAGE), lyophilized powder
Sigma-Aldrich
β-Lactoglobulin B from bovine milk, ≥90% (PAGE)
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥85% (PAGE), lyophilized powder