Merck
Inhibition of stearoyl-CoA desaturase 1 reduces lipogenesis in primary bovine adipocytes.
Lipids (2013-08-10)
A K G Kadegowda, T A Burns, S L Pratt, S K Duckett
ABSTRAKT
The objectives were to determine the effect of stearoyl-CoA desaturase (SCD1) inhibition on adipocyte proliferation, differentiation and cellular lipid metabolism in bovine primary adipocytes. Inhibition of SCD1 activity by sterculic acid (SA) or conjugated linoleic acid, trans-10 cis-12 isomer, (t10, c12-CLA) did not alter adipocyte cellular proliferation, viability or differentiation. In 1,2-[(13)C]-acetate supplemented cells, the mass isotopomer distribution analysis showed that the fractional synthesis rate of [(13)C]-16:0 was reduced (P < 0.01) in SA and t10, c12-CLA treatments compared to control. Of the lipogenic genes, t10, c12-CLA treatment decreased (P < 0.05) the expression of SCD1, acetyl-CoA carboxylase (ACC), fatty acid synthase; whereas SA supplementation decreased (P < 0.05) the expression of ACC. Both SA and t10, c12-CLA increased (P < 0.05) the expression of hormone-sensitive lipase and carnitine palmitoyl transferase involved in lipolysis and oxidation. Inhibition of SCD1 in bovine adipocytes decreases de novo fatty acid synthesis by down-regulating genes involved in lipogenesis and up-regulating genes involved in lipolysis and oxidation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cholesterol Esterase from bovine pancreas, lyophilized powder, ≥200 units/g protein
Sigma-Aldrich
Cholesterol Esterase from Pseudomonas sp., lyophilized powder, ≥200,000 units/g protein
Sigma-Aldrich
Cholesterol Esterase from porcine pancreas, lyophilized, powder, white, ~35 U/mg
Sigma-Aldrich
Cholesterol Esterase from Pseudomonas fluorescens, lyophilized powder, ≥10,000 units/g protein