Merck
  • Lateral growth limitation of corneal fibrils and their lamellar stacking depend on covalent collagen cross-linking by transglutaminase-2 and lysyl oxidases, respectively.
Lateral growth limitation of corneal fibrils and their lamellar stacking depend on covalent collagen cross-linking by transglutaminase-2 and lysyl oxidases, respectively.
The Journal of biological chemistry (2013-11-23)
Lei Wang, Philipp C Uhlig, Eric F Eikenberry, Horst Robenek, Peter Bruckner, Uwe Hansen
ABSTRAKT
Corneal stroma contains an extracellular matrix of orthogonal lamellae formed by parallel and equidistant fibrils with a homogeneous diameter of ~35 nm. This is indispensable for corneal transparency and mechanical functions. However, the mechanisms controlling corneal fibrillogenesis are incompletely understood and the conditions required for lamellar stacking are essentially unknown. Under appropriate conditions, chick embryo corneal fibroblasts can produce an extracellular matrix in vitro resembling primary corneal stroma during embryonic development. Among other requirements, cross-links between fibrillar collagens, introduced by tissue transglutaminase-2, are necessary for the self-assembly of uniform, small diameter fibrils but not their lamellar stacking. By contrast, the subsequent lamellar organization into plywood-like stacks depends on lysyl aldehyde-derived cross-links introduced by lysyl oxidase activity, which, in turn, only weakly influences fibril diameters. These cross-links are introduced at early stages of fibrillogenesis. The enzymes are likely to be important for a correct matrix deposition also during repair of the cornea.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Transglutaminase from guinea pig liver, ≥1.5 units/mg protein, recombinant, expressed in E. coli
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Transglutaminase from guinea pig liver, lyophilized powder, ≥1.5 units/mg protein
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Collagen Type III Antibody, clone IE7-D7, clone 1E7-D7, from mouse
Informacje o cenach i dostępności nie są obecnie dostępne.