Merck
[Blood test for malignant lymphoma].
Nihon rinsho. Japanese journal of clinical medicine (2014-04-15)
Tsutomu Kobayashi, Junya Kuroda, Masafumi Taniwaki
ABSTRAKT
Malignant lymphoma is a neoplastic disease that develops in the lymph system, which consists of various different subtypes. In addition, the differential diagnosis of malignant lymphoma includes infections, autoimmune diseases, allergic diseases, endocrine disorders, and so on. Therefore accurate diagnosis is very important to decide therapeutic strategy. Blood test is the most common examination in clinical practice and used extensively for evaluating etiology, pathology, disease state, efficacy of treatment and disease prognosis of lymphoma. We are required to understand the characteristics of blood examinations correctly and use them appropriately in daily medical practice. Here, we introduce some blood examinations used for treatment of lymphoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein