Merck
Clinical outcomes of AIDS-related Burkitt lymphoma: a multi-institution retrospective survey in Japan.
Japanese journal of clinical oncology (2014-02-22)
Yuki Kojima, Shotaro Hagiwara, Tomoko Uehira, Atsushi Ajisawa, Akira Kitanaka, Junko Tanuma, Seiji Okada, Hirokazu Nagai
ABSTRAKT
Acquired immunodeficiency syndrome-related non-Hodgkin lymphoma is treated similarly to non-acquired immunodeficiency syndrome lymphoma, but it is not clear whether highly intensive regimens are beneficial for acquired immunodeficiency syndrome-related Burkitt lymphoma. We conducted a multicenter retrospective survey to clarify the clinical outcomes of acquired immunodeficiency syndrome-related Burkitt lymphoma in the combined antiretroviral therapy era in Japan. We retrospectively analyzed the outcome of 33 patients with acquired immunodeficiency syndrome-related Burkitt lymphoma, who were diagnosed at five regional hospitals for human immunodeficiency virus/acquired immunodeficiency syndrome in Japan between January 2002 and December 2010. The median follow-up period was 20.0 months (range 0.5-92.7 months). Six (18.2%) patients were treated with cyclophosphamide, vincristine, doxorubicin, high-dose methotrexate, ifosphamide, etoposide and high-dose cytarabine, and 23 (69.7%) patients were treated with hyperfractionated cyclophosphamide, vincristine, doxorubicin, dexamethasone, high-dose methotrexate and high-dose cytarabine. The overall response rate for all patients was 78.8%, with a complete response rate of 72.7%. The two-year overall survival rate was 68.1%. There was no significant difference in overall survival between chemotherapeutic regimens with rituximab (n = 20) and without rituximab (n = 13) (P = 0.49). The two-year overall survival rate was 66.7% for patients receiving cyclophosphamide, vincristine, doxorubicin, dexamethasone, etoposide, ifosfamide and cytarabine, and was 72.6% for patients receiving cyclophosphamide, vincristine, doxorubicin, dexamethasone, methotrexate and cytarabine (P = 0.72). There was one treatment-related death. Highly intensive chemotherapy would bring a high remission rate and prolonged overall survival for patients with acquired immunodeficiency syndrome-related Burkitt lymphoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Prednisone, ≥98%
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ifosfamide, ≥98%
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Ifosfamide, European Pharmacopoeia (EP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Etoposide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture