Merck
  • High-performance thin-layer chromatography analysis of steviol glycosides in Stevia formulations and sugar-free food products, and benchmarking with (ultra) high-performance liquid chromatography.
High-performance thin-layer chromatography analysis of steviol glycosides in Stevia formulations and sugar-free food products, and benchmarking with (ultra) high-performance liquid chromatography.
Journal of chromatography. A (2014-05-31)
Gertrud E Morlock, Stephanie Meyer, Benno F Zimmermann, Jean-Marc Roussel
ABSTRAKT
A high-performance TLC (HPTLC) method was newly developed and validated for analysis of 7 steviol glycosides in 6 different types of food and Stevia formulations. After a minimized one-step sample preparation, 21 samples were developed in parallel, allowing an effective food screening. Depending on the sample application volume, the method was suited to analyze food sample concentrations in the mg/kg range. LOQs of stevioside in natural yoghurt matrix spiked at 0.02, 0.13 and 0.2% were determined by the calibration curve method to be 12ng/band (peak height). ANOVA was successfully passed to prove data homogeneity in the working range (30-600ng/band). The accuracy (recovery tolerance limit, 92-120%), repeatability (3.1-5.4%) and intermediate precision (4.0-8.4%) were determined for stevioside in milk-based matrix including sample preparation and recovery rates at 3 different concentration levels. For the first time, the recording of HPTLC-ESI-MS spectra via the TLC-MS Interface was demonstrated for rebaudioside A. HPTLC contents for rebaudioside A were compared with results of two (U)HPLC methods. The running costs and analysis time of the three different methods were discussed in detail with regard to screening of food products.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
USP
Kwas octowy lodowaty, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Steviol hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
2-Naphthol, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
2-Naphthol, 98%
Sigma-Aldrich
2-Naphthol, 99%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Xylitol, analytical standard
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylitol
Sigma-Aldrich
Xylitol, ≥99% (GC)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
USP
Xylitol, United States Pharmacopeia (USP) Reference Standard
Xylitol, European Pharmacopoeia (EP) Reference Standard