Merck
Intravitreal dexamethasone implant [Ozurdex] for the treatment of nonarteritic anterior ischaemic optic neuropathy.
Documenta ophthalmologica. Advances in ophthalmology (2014-10-03)
Florian Alten, Christoph R Clemens, Peter Heiduschka, Nicole Eter
ABSTRAKT
To evaluate structural and functional outcomes of intravitreal dexamethasone implant (IDI) in three patients presenting with nonarteritic anterior ischaemic optic neuropathy. IDI was administered once in three patients. Best-corrected visual acuity (BCVA), perimetry, volume spectral-domain optical coherence tomography scan of the optic disc (ODV), retinal nerve fibre layer (RNFL) scan and visually evoked potential (VEP) measurements were assessed at baseline and after one and 3 months. Mean BCVA was 20/100 in patient 1 (patient 2: 20/100; patient 3: 20/50) at baseline, 20/60 (patient 2: 20/400) at 1 month and 20/80 (20/400; 20/60) at 3 months. Mean deviation in perimetry developed from -4.90 dB (-22.09 dB; -8.68 dB) to -7.60 dB (-30.75 dB) and -14.23 dB (-30.59 dB; -7.17 dB). ODV and RNFL decreased during follow-up. VEP measurements showed a reduction in amplitudes during the entire observation period. All patients showed a reduction in papilla oedema over time. A functional improvement was not observed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.