Merck
Delivery of negatively charged liposomes into the atherosclerotic plaque of apolipoprotein E-deficient mouse aortic tissue.
Journal of liposome research (2014-01-22)
Siqin Zhaorigetu, Cristian Rodriguez-Aguayo, Anil K Sood, Gabriel Lopez-Berestein, Brian L Walton
ABSTRAKT
Liposomes have been used to diagnose and treat cancer and, to a lesser extent, cardiovascular disease. We previously showed the uptake of anionic liposomes into the atheromas of Watanabe heritable hyperlipidemic rabbits within lipid pools. However, the cellular distribution of anionic liposomes in atherosclerotic plaque remains undescribed. In addition, how anionic liposomes are absorbed into atherosclerotic plaque is unclear. We investigated the uptake and distribution of anionic liposomes in atherosclerotic plaque in aortic tissues from apolipoprotein E-deficient (ApoE(-/-)) mice. To facilitate the tracking of liposomes, we used liposomes containing fluorescently labeled non-silencing small interfering RNA. Confocal microscopy analysis showed the uptake of anionic liposomes into atherosclerotic plaque and colocalization with macrophages. Transmission electron microscopy analysis revealed anionic liposomal accumulation in macrophages. To investigate how anionic liposomes cross the local endothelial barrier, we examined the role of clathrin-mediated endocytosis in human coronary artery endothelial cells (HCAECs) treated with or without the inflammatory cytokine tumor necrosis factor (TNF)-α. Pretreatment with amantadine, an inhibitor of clathrin-mediated endocytosis, significantly decreased liposomal uptake in HCAECs treated with or without TNF-α by 77% and 46%, respectively. Immunoblot analysis showed that endogenous clathrin expression was significantly increased in HCAECs stimulated with TNF-α but was inhibited by amantadine. These studies indicated that clathrin-mediated endocytosis is partly responsible for the uptake of liposomes by endothelial cells. Our results suggest that anionic liposomes target macrophage-rich areas of vulnerable plaque in ApoE(-)(/)(-) mice; this finding may lead to the development of novel diagnostic and therapeutic strategies for treating vulnerable plaque in humans.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
1-Adamantylamine, 97%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture