Merck
Atrazine and its degradates have little effect on the corticosteroid stress response in the zebrafish.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2015-01-28)
Glen Van Der Kraak, Jacquie Matsumoto, Myoungwoo Kim, Alan J Hosmer
ABSTRAKT
The present study examined the effects of atrazine on basal and forced swimming induced changes in whole body cortisol content in adult zebrafish. Zebrafish were exposed to graded concentrations of atrazine or the atrazine degradates deisopropylatrazine (DIA), deethylatrazine (DEA) and diamino-s-chlorotriazine (DACT) for up to 10 days. Some fish were sampled for the measurement of whole body cortisol levels under basal conditions while others were sampled after being subjected to a 20 min swimming challenge in order to quantify stress induced cortisol levels. In one experiment, zebrafish were subjected to two bouts of forced swimming 3h apart to test whether prior atrazine exposure affects the ability of the fish to respond appropriately to a repeated stressor. The results demonstrated that controls not exposed to atrazine and zebrafish exposed to atrazine or the atrazine degradates at nominal concentrations of up to 100 μg/L consistently exhibited increased whole body cortisol content in response to the swimming challenge. Separate analyses revealed few changes in basal or stress induced cortisol levels following atrazine exposure. Overall, these data suggest that atrazine and some of its degradates at the concentrations tested have minimal effects on the cortisol mediated stress response in the zebrafish.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
2-Phenoxyethanol, ≥99%
Supelco
2-Phenoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Phenoxyethanol, analytical standard
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Supelco
Atrazine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Atrazine, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Atrazine, PESTANAL®, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylene glycol monophenyl ether, ≥90%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)