Merck
Brief reports: TRPM7 Senses mechanical stimulation inducing osteogenesis in human bone marrow mesenchymal stem cells.
Stem cells (Dayton, Ohio) (2014-09-30)
E Xiao, H Q Yang, Ye-Hua Gan, Deng-Hui Duan, Lin-Hai He, YunBo Guo, S Q Wang, Yi Zhang
ABSTRAKT
Mesenchymal stem cells (MSCs) are multipotential stem cells residing in the bone marrow. Several studies have shown that mechanical stimulation modulates MSC differentiation through mobilization of second messengers, but the mechanism of mechanotransduction remains poorly understood. In this study, using fluorescence and laser confocal microcopy as well as patch-clamp techniques, we identified the transient receptor potential melastatin type 7 (TRPM7) channel as the key channel involved in mechanotransduction in bone marrow MSCs. TRPM7 knockdown completely abolished the pressure-induced cytosolic Ca(2+) increase and pressure-induced osteogenesis. TRPM7 directly sensed membrane tension, independent of the cytoplasm and the integrity of cytoskeleton. Ca(2+) influx through TRPM7 further triggered Ca(2+) release from the inositol trisphosphate receptor type 2 on the endoplasmic reticulum and promoted NFATc1 nuclear localization and osteogenesis. These results identified a central role of TRPM7 in MSC mechanical stimulation-induced osteogenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
Water, ACS reagent
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Standardowa gęstość 998 kg/m3, H&D Fitzgerald Ltd. Quality
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Deionized
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O