Merck
  • HDL size is more accurate than HDL cholesterol to predict carotid subclinical atherosclerosis in individuals classified as low cardiovascular risk.
HDL size is more accurate than HDL cholesterol to predict carotid subclinical atherosclerosis in individuals classified as low cardiovascular risk.
PloS one (2014-12-04)
Eliane Soler Parra, Natalia Baratella Panzoldo, Vanessa Helena de Souza Zago, Daniel Zanetti Scherrer, Fernanda Alexandre, Jamal Bakkarat, Valeria Sutti Nunes, Edna Regina Nakandakare, Eder Carlos Rocha Quintão, Wilson Nadruz, Eliana Cotta de Faria, Andrei C Sposito
ABSTRAKT
Misclassification of patients as low cardiovascular risk (LCR) remains a major concern and challenges the efficacy of traditional risk markers. Due to its strong association with cholesterol acceptor capacity, high-density lipoprotein (HDL) size has been appointed as a potential risk marker. Hence, we investigate whether HDL size improves the predictive value of HDL-cholesterol in the identification of carotid atherosclerotic burden in individuals stratified to be at LCR. 284 individuals (40-75 years) classified as LCR by the current US guidelines were selected in a three-step procedure from primary care centers of the cities of Campinas and Americana, SP, Brazil. Apolipoprotein B-containing lipoproteins were precipitated by polyethylene glycol and HDL size was measured by dynamic light scattering (DLS) technique. Participants were classified in tertiles of HDL size (<7.57; 7.57-8.22; >8.22 nm). Carotid intima-media thickness (cIMT) <0.90 mm (80th percentile) was determined by high resolution ultrasonography and multivariate ordinal regression models were used to assess the association between cIMT across HDL size and levels of lipid parameters. HDL-cholesterol was not associated with cIMT. In contrast, HDL size >8.22 nm was independently associated with low cIMT in either unadjusted and adjusted models for age, gender and Homeostasis Model Assessment 2 index for insulin sensitivity, ethnicity and body mass index (Odds ratio 0.23; 95% confidence interval 0.07-0.74, p = 0.013). The mean HDL size estimated with DLS constitutes a better predictor for subclinical carotid atherosclerosis than the conventional measurements of plasma HDL-cholesterol in individuals classified as LCR.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Paraoxon-ethyl, PESTANAL®, analytical standard
Sigma-Aldrich
Paraoxon-ethyl, ≥90%, oil