Merck
Cyclooxygenase-2 regulates NLRP3 inflammasome-derived IL-1β production.
Journal of cellular physiology (2014-10-09)
Kuo-Feng Hua, Ju-Ching Chou, Shuk-Man Ka, Yu-Ling Tasi, Ann Chen, Shih-Hsiung Wu, Hsiao-Wen Chiu, Wei-Ting Wong, Yih-Fuh Wang, Change-Ling Tsai, Chen-Lung Ho, Cheng-Hsiu Lin
ABSTRAKT
The NLR family, pyrin domain-containing 3 (NLRP3) inflammasome is a reactive oxygen species-sensitive multiprotein complex that regulates IL-1β maturation via caspase-1. It also plays an important role in the pathogenesis of inflammation-related disease. Cyclooxygenase-2 (COX-2) is induced by inflammatory stimuli and contributes to the pathogenesis of inflammation-related diseases. However, there is currently little known about the relationship between COX-2 and the NLRP3 inflammasome. Here, we describe a novel role for COX-2 in regulating the activation of the NLRP3 inflammasome. NLRP3 inflammasome-derived IL-1β secretion and pyroptosis in macrophages were reduced by pharmaceutical inhibition or genetic knockdown of COX-2. COX-2 catalyzes the synthesis of prostaglandin E2 and increases IL-1β secretion. Conversely, pharmaceutical inhibition or genetic knockdown of prostaglandin E2 receptor 3 reduced IL-1β secretion. The underlying mechanisms for the COX-2-mediated increase in NLRP3 inflammasome activation were determined to be the following: (1) enhancement of lipopolysaccharide-induced proIL-1β and NLRP3 expression by increasing NF-κB activation and (2) enhancement of the caspase-1 activation by increasing damaged mitochondria, mitochondrial reactive oxygen species production and release of mitochondrial DNA into cytosol. Furthermore, inhibition of COX-2 in mice in vivo with celecoxib reduced serum levels of IL-1β and caspase-1 activity in the spleen and liver in response to lipopolysaccharide (LPS) challenge. These findings provide new insights into how COX-2 regulates the activation of the NLRP3 inflammasome and suggest that it may be a new potential therapeutic target in NLRP3 inflammasome-related diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Celecoxib, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
SAFC
L-Glutamine
Sigma-Aldrich
AH6809, ≥98%, crystalline solid or supercooled liquid
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-798106, ≥98% (HPLC)